The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006802.3 | NP_006793.1 | 4 - 5 | 453 | 112 |
GenBank mRNA | AK128438.1 | - | 4 - 5 | 1262 | 112 |
AK222554.1 | - | 4 - 5 | 329 | 112 | |
AK291181.1 | - | 4 - 5 | 334 | 112 | |
AK292116.1 | - | 4 - 5 | 364 | 112 | |
BC002395.2 | - | 4 - 5 | 346 | 112 | |
BC011523.1 | - | 4 - 5 | 410 | 112 | |
DA835019.1 | - | 4 - 5 | 453 | 112 | |
U08815.1 | - | 4 - 5 | 316 | 112 |