The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320830.1 | NP_001307759.1 | 6 - 7 | 677 | 61 |
NM_006802.3 | NP_006793.1 | 8 - 9 | 836 | 61 | |
GenBank mRNA | AK128438.1 | - | 8 - 9 | 1645 | 61 |
AK222554.1 | - | 8 - 9 | 712 | 61 | |
AK291181.1 | - | 8 - 9 | 717 | 61 | |
AK292116.1 | - | 8 - 9 | 747 | 61 | |
AK301424.1 | - | 6 - 7 | 554 | 61 | |
BC002395.2 | - | 8 - 9 | 729 | 61 | |
BC011523.1 | - | 8 - 9 | 793 | 61 | |
U08815.1 | - | 8 - 9 | 699 | 61 |