Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ST3GAL4
Entrez Gene ID
6484
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ
Chromosome Location
Chr.11: 126355645 - 126459601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254757.1 NP_001241686.1 5 - 6 531 87
NM_001254758.1 NP_001241687.1 5 - 6 404 87
NM_001254759.1 NP_001241688.1 3 - 4 548 87
NM_006278.2 NP_006269.1 5 - 6 730 87
XM_011542959.1 XP_011541261.1 4 - 5 333 87
XM_011542960.2 XP_011541262.1 4 - 5 333 87
XM_017018134.1 XP_016873623.1 5 - 6 416 87
GenBank mRNA AF035249.1 - 1 - 2 100 87
AF516602.1 - 3 - 4 258 87
AF516603.1 - 3 - 4 228 87
AF516604.1 - 3 - 4 258 87
AF525084.1 - 3 - 4 288 87
AK021929.1 - 3 - 4 232 87
AK291577.1 - 4 - 5 360 87
AK314738.1 - 4 - 5 330 87
AL520639.3 - 5 - 6 489 87
AL529887.3 - 5 - 6 405 87
AY040826.1 - 4 - 5 291 87
BC010645.1 - 5 - 6 404 87
BI259855.1 - 5 - 6 499 87
CR456858.1 - 4 - 5 261 87
L23767.1 - 3 - 4 270 87
X74570.1 - 5 - 6 423 87

View Full Product Details