Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
Chromosome Location
Chr.11: 130069821 - 130144811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142276.1 NP_001135748.1 15 - 16 2155 88
NM_001142277.1 NP_001135749.1 14 - 15 1987 88
NM_001142278.1 NP_001135750.1 11 - 12 1468 88
NM_001243299.1 NP_001230228.1 15 - 16 2073 88
NM_001642.2 NP_001633.1 16 - 17 2191 88
NR_024515.1 12 - 13 1653 88
NR_024516.1 13 - 14 1725 88
GenBank mRNA AF168956.1 - 16 - 17 2037 88
AK056883.1 - 12 - 13 1580 88
AK122834.1 - 1 - 2 2425 88
AK124457.1 - 1 - 2 1722 88
AK128162.1 - 15 - 16 2066 88
AK225283.1 - 14 - 15 1914 88
AK296765.1 - 9 - 10 1017 88
AK300378.1 - 13 - 14 1652 88
AK304736.1 - 14 - 15 1896 88
AK308248.1 - 13 - 14 1699 88
AK308932.1 - 3 - 4 247 88
BC000373.2 - 15 - 16 2055 88
BC004371.1 - 11 - 12 1373 88
BX647107.1 - 1 - 2 3697 88
L09209.1 - 16 - 17 2106 88
L19597.1 - 14 - 15 1898 88
L23113.1 - 11 - 12 1316 88
L23114.1 - 14 - 15 1754 88
L27631.1 - 16 - 17 2107 88
S60099.1 - 16 - 17 2106 88
Z22572.1 - 16 - 17 2101 88

View Full Product Details