The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142276.1 | NP_001135748.1 | 16 - 17 | 2311 | 76 |
NM_001142277.1 | NP_001135749.1 | 15 - 16 | 2143 | 76 | |
NM_001142278.1 | NP_001135750.1 | 12 - 13 | 1624 | 76 | |
NM_001243299.1 | NP_001230228.1 | 16 - 17 | 2229 | 76 | |
NM_001642.2 | NP_001633.1 | 17 - 18 | 2347 | 76 | |
NR_024515.1 | 13 - 14 | 1809 | 76 | ||
NR_024516.1 | 14 - 15 | 1881 | 76 | ||
GenBank mRNA | AF168956.1 | - | 17 - 18 | 2193 | 76 |
AK056883.1 | - | 13 - 14 | 1736 | 76 | |
AK122834.1 | - | 2 - 3 | 2581 | 76 | |
AK124457.1 | - | 2 - 3 | 1878 | 76 | |
AK128162.1 | - | 16 - 17 | 2222 | 76 | |
AK225283.1 | - | 15 - 16 | 2070 | 76 | |
AK296765.1 | - | 10 - 11 | 1173 | 76 | |
AK300378.1 | - | 14 - 15 | 1808 | 76 | |
AK304736.1 | - | 15 - 16 | 2052 | 76 | |
AK308248.1 | - | 14 - 15 | 1855 | 76 | |
AK308932.1 | - | 4 - 5 | 403 | 76 | |
BC000373.2 | - | 16 - 17 | 2211 | 76 | |
BC004371.1 | - | 12 - 13 | 1529 | 76 | |
BX647107.1 | - | 2 - 3 | 3853 | 76 | |
L09209.1 | - | 17 - 18 | 2262 | 76 | |
L19597.1 | - | 15 - 16 | 2054 | 76 | |
L23113.1 | - | 12 - 13 | 1472 | 76 | |
L23114.1 | - | 15 - 16 | 1910 | 76 | |
L27631.1 | - | 17 - 18 | 2263 | 76 | |
S60099.1 | - | 17 - 18 | 2262 | 76 | |
Z22572.1 | - | 17 - 18 | 2257 | 76 |