Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
Chromosome Location
Chr.11: 130069821 - 130144811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142276.1 NP_001135748.1 2 - 3 429 95
NM_001142277.1 NP_001135749.1 2 - 3 429 95
NM_001142278.1 NP_001135750.1 2 - 3 429 95
NM_001243299.1 NP_001230228.1 2 - 3 347 95
NM_001642.2 NP_001633.1 2 - 3 429 95
GenBank mRNA AF168956.1 - 2 - 3 275 95
AK225283.1 - 2 - 3 356 95
AK296765.1 - 2 - 3 356 95
AK304736.1 - 2 - 3 338 95
BC000373.2 - 2 - 3 329 95
BC004371.1 - 2 - 3 334 95
DC336210.1 - 2 - 3 429 95
L09209.1 - 2 - 3 344 95
L19597.1 - 2 - 3 340 95
L23114.1 - 1 - 2 160 95
L27631.1 - 2 - 3 345 95
S60099.1 - 2 - 3 344 95
Z22572.1 - 2 - 3 339 95

View Full Product Details