The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281748.1 | NP_001268677.1 | 2 - 3 | 385 | 146 |
NM_001281749.1 | NP_001268678.1 | 2 - 3 | 385 | 146 | |
NM_025164.4 | NP_079440.3 | 2 - 3 | 385 | 146 | |
XM_005271481.2 | XP_005271538.1 | 2 - 3 | 423 | 146 | |
XM_005271482.4 | XP_005271539.2 | 2 - 3 | 423 | 146 | |
XM_005271484.3 | XP_005271541.2 | 2 - 3 | 423 | 146 | |
XM_011542721.2 | XP_011541023.1 | 2 - 3 | 385 | 146 | |
XM_011542722.1 | XP_011541024.1 | 2 - 3 | 423 | 146 | |
XM_011542726.1 | XP_011541028.1 | 2 - 3 | 423 | 146 | |
XM_017017424.1 | XP_016872913.1 | 2 - 3 | 423 | 146 | |
GenBank mRNA | BC121188.1 | - | 2 - 3 | 278 | 146 |
BC128510.1 | - | 2 - 3 | 247 | 146 | |
BC128511.1 | - | 2 - 3 | 241 | 146 |