Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100129291
- Entrez Gene ID
-
100129291
- Gene Name
- chromosome X open reading frame 49 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 71662991 - 71666868 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_136576.1 |
|
5 - 6 |
1639 |
79 |
GenBank mRNA |
BG201338.1 |
- |
3 - 4 |
172 |
79 |
- Gene Symbol
- CXorf49
- Entrez Gene ID
-
100130361
- Gene Name
- chromosome X open reading frame 49
- Gene Aliases
-
CXorf49B
- Chromosome Location
-
Chr.X: 71714374 - 71718285 on Build GRCh38
- Gene Symbol
- CXorf49B
- Entrez Gene ID
-
100132994
- Gene Name
- chromosome X open reading frame 49B
- Gene Aliases
-
CXorf49
- Chromosome Location
-
Chr.X: 71763290 - 71767201 on Build GRCh38
View Full Product Details