Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED25
Entrez Gene ID
81857
Gene Name
mediator complex subunit 25
Gene Aliases
ACID1, ARC92, BVSYS, CMT2B2, P78, PTOV2, TCBAP0758
Chromosome Location
Chr.19: 49818279 - 49840384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030973.3 NP_112235.2 11 - 12 1377 126
XM_011527353.1 XP_011525655.1 11 - 12 1369 126
GenBank mRNA AF261072.1 - 11 - 12 1356 126
AF283769.2 - 6 - 7 837 126
AJ617479.1 - 11 - 12 1354 126
AK055844.1 - 11 - 12 1367 126
AK289460.1 - 11 - 12 1367 126
AK298897.1 - 11 - 12 1367 126
AL136746.1 - 11 - 12 1354 126
AY533507.1 - 11 - 12 1314 126
BC021543.1 - 10 - 11 1157 126
BC024312.2 - 7 - 8 803 126
BC065297.1 - 11 - 12 1350 126
EU392500.1 - 6 - 7 675 126
EU392501.1 - 5 - 6 519 126
EU392519.1 - 4 - 5 253 126

View Full Product Details