The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207408.1 | NP_997291.1 | 1 - 2 | 93 | 125 |
GenBank mRNA | AK131015.1 | - | 1 - 2 | 93 | 125 |
BC130495.1 | - | 1 - 2 | 101 | 125 | |
BC130497.1 | - | 1 - 2 | 101 | 125 |