The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270523.1 | NP_001257452.1 | 1 - 2 | 104 | 97 |
NM_021728.3 | NP_068374.1 | 1 - 2 | 104 | 97 | |
NR_073034.1 | 1 - 2 | 104 | 97 | ||
GenBank mRNA | AB593058.1 | - | 1 - 2 | 104 | 97 |
BC032579.1 | - | 1 - 2 | 105 | 97 | |
CV812961.1 | - | 1 - 2 | 81 | 97 | |
DB294481.1 | - | 1 - 2 | 107 | 97 |