The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270523.1 | NP_001257452.1 | 2 - 3 | 160 | 109 |
NM_021728.3 | NP_068374.1 | 2 - 3 | 160 | 109 | |
NR_073034.1 | 2 - 3 | 160 | 109 | ||
GenBank mRNA | AB593058.1 | - | 2 - 3 | 160 | 109 |
BC032579.1 | - | 2 - 3 | 161 | 109 | |
CV812961.1 | - | 2 - 3 | 137 | 109 | |
DB294481.1 | - | 2 - 3 | 163 | 109 |