The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323553.1 | NP_001310482.1 | 3 - 4 | 505 | 62 |
NM_001323554.1 | NP_001310483.1 | 3 - 4 | 1444 | 62 | |
NM_001323555.1 | NP_001310484.1 | 3 - 4 | 1513 | 62 | |
NM_001323556.1 | NP_001310485.1 | 3 - 4 | 286 | 62 | |
NM_001323557.1 | NP_001310486.1 | 3 - 4 | 355 | 62 | |
NM_001323558.1 | NP_001310487.1 | 3 - 4 | 436 | 62 | |
NM_015137.5 | NP_055952.2 | 3 - 4 | 436 | 62 | |
NR_136615.1 | 3 - 4 | 505 | 62 | ||
NR_136616.1 | 3 - 4 | 436 | 62 | ||
GenBank mRNA | BC071611.1 | - | 3 - 4 | 413 | 62 |
BC108667.1 | - | 3 - 4 | 404 | 62 | |
BC152442.1 | - | 3 - 4 | 404 | 62 | |
CR749243.1 | - | 3 - 4 | 246 | 62 | |
D63477.1 | - | 3 - 4 | 404 | 62 | |
DB451954.2 | - | 3 - 4 | 415 | 62 | |
DB483230.1 | - | 3 - 4 | 436 | 62 | |
DB727938.1 | - | 3 - 4 | 484 | 62 | |
HY095205.1 | - | 3 - 4 | 432 | 62 | |
HY124760.1 | - | 5 - 6 | 432 | 62 |