The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168551.1 | NP_001162023.1 | 14 - 15 | 1805 | 106 |
NM_018087.4 | NP_060557.3 | 14 - 15 | 1925 | 106 | |
NR_033142.1 | 13 - 14 | 1804 | 106 | ||
XM_011541766.2 | XP_011540068.1 | 14 - 15 | 1767 | 106 | |
GenBank mRNA | AK001269.1 | - | 14 - 15 | 1808 | 106 |
AK022618.1 | - | 2 - 3 | 81 | 106 | |
AK091439.1 | - | 8 - 9 | 611 | 106 | |
AK295000.1 | - | 14 - 15 | 1589 | 106 | |
AK298909.1 | - | 13 - 14 | 1548 | 106 | |
AK302910.1 | - | 12 - 13 | 1383 | 106 | |
AL354613.1 | - | 14 - 15 | 1805 | 106 | |
BC003082.2 | - | 14 - 15 | 1875 | 106 | |
DQ141696.1 | - | 14 - 15 | 1634 | 106 |