Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEPDC1B
Entrez Gene ID
55789
Gene Name
DEP domain containing 1B
Gene Aliases
BRCC3, XTP1
Chromosome Location
Chr.5: 60596912 - 60700166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145208.1 NP_001138680.1 2 - 3 379 148
NM_018369.2 NP_060839.2 2 - 3 379 148
XM_011543509.2 XP_011541811.1 2 - 3 372 148
XM_011543510.2 XP_011541812.1 2 - 3 347 148
XM_017009629.1 XP_016865118.1 2 - 3 347 148
GenBank mRNA AF303178.1 - 2 - 3 347 148
AF490257.1 - 2 - 3 306 148
AK002114.1 - 2 - 3 347 148
AK290684.1 - 2 - 3 374 148
AK299242.1 - 3 - 4 413 148
AK300782.1 - 2 - 3 347 148
BC019075.1 - 2 - 3 342 148
DC299445.1 - 2 - 3 380 148

View Full Product Details