The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152731.2 | NP_689944.2 | 4 - 5 | 1103 | 81 |
XM_005248889.1 | XP_005248946.1 | 2 - 3 | 271 | 81 | |
XM_017010406.1 | XP_016865895.1 | 4 - 5 | 654 | 81 | |
XM_017010407.1 | XP_016865896.1 | 4 - 5 | 637 | 81 | |
XM_017010408.1 | XP_016865897.1 | 5 - 6 | 1184 | 81 | |
GenBank mRNA | AK303705.1 | - | 3 - 4 | 473 | 81 |
AK315926.1 | - | 1 - 2 | 67 | 81 | |
BC022988.1 | - | 3 - 4 | 498 | 81 | |
DC411761.1 | - | 3 - 4 | 473 | 81 |