The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152731.2 | NP_689944.2 | 5 - 6 | 1294 | 146 |
XM_005248889.1 | XP_005248946.1 | 3 - 4 | 462 | 146 | |
XM_017010406.1 | XP_016865895.1 | 5 - 6 | 845 | 146 | |
XM_017010407.1 | XP_016865896.1 | 5 - 6 | 828 | 146 | |
XM_017010408.1 | XP_016865897.1 | 6 - 7 | 1375 | 146 | |
GenBank mRNA | AK054724.1 | - | 6 - 7 | 876 | 146 |
AK303705.1 | - | 4 - 5 | 664 | 146 | |
AK315926.1 | - | 2 - 3 | 258 | 146 | |
BC022988.1 | - | 4 - 5 | 689 | 146 |