Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO16
Entrez Gene ID
157574
Gene Name
F-box protein 16
Gene Aliases
FBX16
Chromosome Location
Chr.8: 28428408 - 28490318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258211.1 NP_001245140.1 1 - 2 129 68
NM_172366.3 NP_758954.1 1 - 2 129 68
GenBank mRNA AF453435.1 - 1 - 2 38 68
AL560247.3 - 1 - 2 89 68
BC102026.1 - 1 - 2 48 68
BC102027.1 - 1 - 2 48 68
BC102028.1 - 1 - 2 48 68
CB134505.1 - 1 - 2 131 68

View Full Product Details