Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AURKC
- Entrez Gene ID
-
6795
- Gene Name
- aurora kinase C
- Gene Aliases
-
AIE2, AIK3, ARK3, AurC, HEL-S-90, SPGF5, STK13, aurora-C
- Chromosome Location
-
Chr.19: 57230317 - 57235550 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003160.2 |
NP_003151.2 |
1 - 2 |
114 |
64 |
View Full Product Details