The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005924.4 | NP_005915.2 | 2 - 3 | 965 | 72 |
GenBank mRNA | AK309472.1 | - | 4 - 5 | 594 | 72 |
AK313386.1 | - | 2 - 3 | 890 | 72 | |
BC017021.1 | - | 2 - 3 | 862 | 72 | |
CR536553.1 | - | 2 - 3 | 681 | 72 | |
X82629.1 | - | 2 - 3 | 946 | 72 |