Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR2C1
Entrez Gene ID
7181
Gene Name
nuclear receptor subfamily 2 group C member 1
Gene Aliases
TR2
Chromosome Location
Chr.12: 95020229 - 95073628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003297.3 NP_003288.2 12 - 13 1782 138
XM_005269133.2 XP_005269190.1 12 - 13 1754 138
XM_006719585.2 XP_006719648.1 13 - 14 1807 138
XM_011538717.2 XP_011537019.1 10 - 11 1481 138
GenBank mRNA AK291319.1 - 12 - 13 1746 138
BC026074.1 - 9 - 10 1882 138
JQ308177.1 - 11 - 12 1526 138
M29960.1 - 12 - 13 1582 138

View Full Product Details