The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316973.1 | NP_001303902.1 | 2 - 3 | 237 | 114 |
NM_001316975.1 | NP_001303904.1 | 2 - 3 | 237 | 114 | |
NM_001316976.1 | NP_001303905.1 | 2 - 3 | 237 | 114 | |
NM_032368.4 | NP_115744.2 | 2 - 3 | 337 | 114 | |
XM_005263506.2 | XP_005263563.1 | 2 - 3 | 232 | 114 | |
XM_005263509.2 | XP_005263566.1 | 2 - 3 | 232 | 114 | |
XM_017002546.1 | XP_016858035.1 | 2 - 3 | 212 | 114 | |
XM_017002547.1 | XP_016858036.1 | 2 - 3 | 232 | 114 | |
GenBank mRNA | AB060688.1 | - | 2 - 3 | 189 | 114 |
AK312550.1 | - | 2 - 3 | 179 | 114 | |
BC007240.1 | - | 2 - 3 | 239 | 114 | |
BC063443.1 | - | 1 - 2 | 144 | 114 | |
BI772110.1 | - | 2 - 3 | 194 | 114 | |
DA538145.1 | - | 2 - 3 | 237 | 114 | |
DB106543.1 | - | 1 - 2 | 169 | 114 |