Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LZIC
Entrez Gene ID
84328
Gene Name
leucine zipper and CTNNBIP1 domain containing
Gene Aliases
-
Chromosome Location
Chr.1: 9922113 - 9943427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316973.1 NP_001303902.1 4 - 5 480 131
NM_001316974.1 NP_001303903.1 4 - 5 423 131
NM_001316975.1 NP_001303904.1 4 - 5 480 131
NM_001316976.1 NP_001303905.1 4 - 5 480 131
NM_032368.4 NP_115744.2 4 - 5 580 131
XM_005263506.2 XP_005263563.1 4 - 5 475 131
XM_005263509.2 XP_005263566.1 4 - 5 475 131
XM_017002546.1 XP_016858035.1 4 - 5 455 131
XM_017002547.1 XP_016858036.1 4 - 5 475 131
GenBank mRNA AB060688.1 - 4 - 5 432 131
AK304348.1 - 4 - 5 360 131
AK312550.1 - 4 - 5 422 131
BC007240.1 - 4 - 5 482 131
BC063443.1 - 3 - 4 387 131
BI772110.1 - 4 - 5 437 131
DA538145.1 - 4 - 5 480 131
DB106543.1 - 3 - 4 412 131

View Full Product Details