The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316973.1 | NP_001303902.1 | 5 - 6 | 581 | 94 |
NM_001316974.1 | NP_001303903.1 | 5 - 6 | 524 | 94 | |
NM_001316975.1 | NP_001303904.1 | 5 - 6 | 581 | 94 | |
NM_032368.4 | NP_115744.2 | 5 - 6 | 681 | 94 | |
XM_005263506.2 | XP_005263563.1 | 5 - 6 | 576 | 94 | |
XM_017002546.1 | XP_016858035.1 | 5 - 6 | 556 | 94 | |
XM_017002547.1 | XP_016858036.1 | 5 - 6 | 576 | 94 | |
GenBank mRNA | AB060688.1 | - | 5 - 6 | 533 | 94 |
AK304348.1 | - | 5 - 6 | 461 | 94 | |
AK312550.1 | - | 5 - 6 | 523 | 94 | |
BC007240.1 | - | 5 - 6 | 583 | 94 | |
BC063443.1 | - | 4 - 5 | 488 | 94 | |
BI772110.1 | - | 5 - 6 | 538 | 94 | |
BQ018333.1 | - | 3 - 4 | 94 |