The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008225.2 | NP_001008226.1 | 4 - 5 | 790 | 140 |
NM_001190847.1 | NP_001177776.1 | 4 - 5 | 790 | 140 | |
NM_001190848.1 | NP_001177777.1 | 4 - 5 | 790 | 140 | |
NM_001190849.1 | NP_001177778.1 | 4 - 5 | 790 | 140 | |
NM_001190850.1 | NP_001177779.1 | 4 - 5 | 790 | 140 | |
NM_013316.3 | NP_037448.2 | 4 - 5 | 790 | 140 | |
XM_017012235.1 | XP_016867724.1 | 4 - 5 | 651 | 140 | |
GenBank mRNA | AF180475.1 | - | 3 - 4 | 459 | 140 |
AK074493.1 | - | 4 - 5 | 756 | 140 | |
AK300365.1 | - | 4 - 5 | 756 | 140 | |
AK314540.1 | - | 4 - 5 | 755 | 140 | |
AL389980.1 | - | 4 - 5 | 744 | 140 | |
BC035590.1 | - | 4 - 5 | 750 | 140 | |
U71267.1 | - | 4 - 5 | 740 | 140 | |
U71268.1 | - | 4 - 5 | 740 | 140 |