Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM5
Entrez Gene ID
2915
Gene Name
glutamate metabotropic receptor 5
Gene Aliases
GPRC1E, MGLUR5, PPP1R86, mGlu5
Chromosome Location
Chr.11: 88504576 - 89065971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000842.4 NP_000833.1 8 - 9 3024 77
XM_017017627.1 XP_016873116.1 8 - 9 3022 77
GenBank mRNA AK291362.1 - 8 - 9 2994 77
D28538.1 - 7 - 8 2776 77
FJ588506.1 - 7 - 8 2626 77
FJ668670.1 - 7 - 8 2626 77

View Full Product Details