Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
Chromosome Location
Chr.9: 35073838 - 35080016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004629.1 NP_004620.1 6 - 7 1267 91
GenBank mRNA AJ007669.1 - 6 - 7 1267 91
AK293427.1 - 7 - 8 957 91
AK311348.1 - 5 - 6 741 91
AK312987.1 - 6 - 7 1192 91
BC000032.2 - 6 - 7 913 91
BC011623.2 - 6 - 7 1040 91
BT009813.1 - 6 - 7 775 91
KU159422.1 - 6 - 7 1298 91
KU178053.1 - 6 - 7 775 91
KU178054.1 - 6 - 7 775 91
KU178055.1 - 6 - 7 775 91
KU178056.1 - 3 - 4 349 91
U70310.1 - 6 - 7 909 91

View Full Product Details