Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUSC2
Entrez Gene ID
9853
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
Iporin
Chromosome Location
Chr.9: 35487999 - 35561898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135999.1 NP_001129471.1 11 - 12 4918 74
NM_014806.3 NP_055621.1 11 - 12 4662 74
NR_052015.2 11 - 12 4337 74
XM_006716896.2 XP_006716959.1 10 - 11 4509 74
GenBank mRNA AB002373.2 - 11 - 12 4504 74
BC029647.1 - 2 - 3 477 74
BC064843.1 - 10 - 11 3706 74
BC082245.1 - 2 - 3 552 74
BC132766.1 - 10 - 11 4447 74
BC132770.1 - 10 - 11 4447 74
BC146654.1 - 11 - 12 4636 74
BC150211.1 - 11 - 12 4249 74
BC150262.1 - 11 - 12 4515 74

View Full Product Details