Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC8A2
Entrez Gene ID
6543
Gene Name
solute carrier family 8 member A2
Gene Aliases
NCX2
Chromosome Location
Chr.19: 47428017 - 47472050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015063.2 NP_055878.1 7 - 8 2287 77
XM_005259172.1 XP_005259229.1 6 - 7 2078 77
XM_017027159.1 XP_016882648.1 4 - 5 435 77
GenBank mRNA AB029010.1 - 7 - 8 2102 77
AK293836.1 - 5 - 6 500 77
AK293860.1 - 6 - 7 1420 77
AK302552.1 - 7 - 8 2287 77
AL512764.1 - 6 - 7 1397 77

View Full Product Details