The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015063.2 | NP_055878.1 | 8 - 9 | 2388 | 119 |
XM_005259172.1 | XP_005259229.1 | 7 - 8 | 2179 | 119 | |
XM_017027159.1 | XP_016882648.1 | 5 - 6 | 536 | 119 | |
GenBank mRNA | AB029010.1 | - | 8 - 9 | 2203 | 119 |
AK293457.1 | - | 8 - 9 | 1129 | 119 | |
AK293836.1 | - | 6 - 7 | 601 | 119 | |
AK293860.1 | - | 7 - 8 | 1521 | 119 | |
AK302552.1 | - | 8 - 9 | 2388 | 119 | |
AL512764.1 | - | 7 - 8 | 1498 | 119 |