The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166212.1 | NP_001159684.1 | 2 - 3 | 297 | 81 |
NM_013246.2 | NP_037378.1 | 2 - 3 | 378 | 81 | |
GenBank mRNA | AF172854.1 | - | 2 - 3 | 227 | 81 |
AF176911.1 | - | 2 - 3 | 271 | 81 | |
AK298052.1 | - | 2 - 3 | 297 | 81 | |
AY049779.1 | - | 2 - 3 | 184 | 81 | |
BC012939.1 | - | 2 - 3 | 254 | 81 | |
BC066229.1 | - | 2 - 3 | 186 | 81 | |
BC066230.1 | - | 2 - 3 | 186 | 81 | |
BC066231.1 | - | 2 - 3 | 186 | 81 | |
BM846622.1 | - | 2 - 3 | 380 | 81 | |
DC393345.1 | - | 2 - 3 | 297 | 81 |