The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318934.1 | NP_001305863.1 | 19 - 20 | 2411 | 137 |
NM_001318935.1 | NP_001305864.1 | 19 - 20 | 2414 | 137 | |
NM_003071.3 | NP_003062.2 | 19 - 20 | 2414 | 137 | |
NM_139048.2 | NP_620636.1 | 19 - 20 | 2414 | 137 | |
XM_017007079.1 | XP_016862568.1 | 19 - 20 | 2411 | 137 | |
GenBank mRNA | AB209052.1 | - | 19 - 20 | 2196 | 137 |
AK291231.1 | - | 19 - 20 | 2361 | 137 | |
BC044659.1 | - | 19 - 20 | 2379 | 137 | |
L34673.1 | - | 19 - 20 | 2373 | 137 |