The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318934.1 | NP_001305863.1 | 22 - 23 | 2836 | 149 |
NM_001318935.1 | NP_001305864.1 | 22 - 23 | 2839 | 149 | |
NM_003071.3 | NP_003062.2 | 22 - 23 | 2839 | 149 | |
NM_139048.2 | NP_620636.1 | 22 - 23 | 2839 | 149 | |
XM_011513091.1 | XP_011511393.1 | 22 - 23 | 2851 | 149 | |
XM_011513092.1 | XP_011511394.1 | 22 - 23 | 2851 | 149 | |
XM_011513093.1 | XP_011511395.1 | 22 - 23 | 2848 | 149 | |
XM_017007078.1 | XP_016862567.1 | 22 - 23 | 2827 | 149 | |
XM_017007079.1 | XP_016862568.1 | 22 - 23 | 2836 | 149 | |
GenBank mRNA | AB209052.1 | - | 22 - 23 | 2621 | 149 |
AK291231.1 | - | 22 - 23 | 2786 | 149 | |
BC044659.1 | - | 22 - 23 | 2804 | 149 | |
L34673.1 | - | 22 - 23 | 2798 | 149 |