Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCOLCE2
Entrez Gene ID
26577
Gene Name
procollagen C-endopeptidase enhancer 2
Gene Aliases
PCPE2
Chromosome Location
Chr.3: 142817860 - 142889203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013363.3 NP_037495.1 5 - 6 1018 136
GenBank mRNA AF098269.1 - 5 - 6 744 136
AK075478.1 - 5 - 6 1015 136
AK223633.1 - 5 - 6 940 136
AK315121.1 - 5 - 6 786 136
AY035400.1 - 5 - 6 907 136
AY358557.1 - 5 - 6 1017 136
BC006265.2 - 5 - 6 771 136
CR536570.1 - 5 - 6 711 136

View Full Product Details