The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005224.2 | NP_005215.1 | 7 - 8 | 1783 | 55 |
XM_005259513.4 | XP_005259570.1 | 7 - 8 | 2239 | 55 | |
XM_005259514.4 | XP_005259571.1 | 7 - 8 | 1922 | 55 | |
XM_017026445.1 | XP_016881934.1 | 7 - 8 | 1948 | 55 | |
XM_017026446.1 | XP_016881935.1 | 5 - 6 | 991 | 55 | |
XM_017026447.1 | XP_016881936.1 | 5 - 6 | 831 | 55 | |
GenBank mRNA | AY152547.1 | - | 6 - 7 | 1693 | 55 |
BC033163.1 | - | 5 - 6 | 1043 | 55 | |
BC060828.1 | - | 7 - 8 | 1783 | 55 | |
U88047.1 | - | 6 - 7 | 1693 | 55 |