Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207025.2 NP_001193954.1 1 - 2 85 81
NM_001207026.1 NP_001193955.1 1 - 2 85 81
NM_001247994.1 NP_001234923.1 1 - 2 85 81
NM_002698.4 NP_002689.1 1 - 2 85 81
XM_005259010.3 XP_005259067.1 1 - 2 102 81
XM_011527040.2 XP_011525342.1 1 - 2 102 81
XM_011527041.2 XP_011525343.1 1 - 2 102 81
XM_011527042.2 XP_011525344.1 1 - 2 102 81
XM_011527043.2 XP_011525345.2 1 - 2 102 81
XM_017026884.1 XP_016882373.1 1 - 2 103 81
XM_017026891.1 XP_016882380.1 1 - 2 103 81
XM_017026892.1 XP_016882381.1 1 - 2 103 81
XM_017026893.1 XP_016882382.1 1 - 2 102 81
XM_017026894.1 XP_016882383.1 1 - 2 102 81
XM_017026896.1 XP_016882385.1 1 - 2 103 81
GenBank mRNA BC006101.1 - 1 - 2 77 81
CB995118.1 - 1 - 2 118 81
M36542.1 - 1 - 2 77 81
M36653.1 - 1 - 2 89 81
X13810.1 - 1 - 2 77 81
X53468.1 - 1 - 2 89 81

View Full Product Details