Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207025.2 NP_001193954.1 9 - 10 866 59
NM_001207026.1 NP_001193955.1 9 - 10 866 59
NM_001247994.1 NP_001234923.1 9 - 10 818 59
NM_002698.4 NP_002689.1 9 - 10 818 59
XM_005259010.3 XP_005259067.1 9 - 10 835 59
XM_011527040.2 XP_011525342.1 9 - 10 949 59
XM_011527041.2 XP_011525343.1 9 - 10 949 59
XM_011527042.2 XP_011525344.1 9 - 10 901 59
XM_011527043.2 XP_011525345.2 10 - 11 1246 59
XM_017026884.1 XP_016882373.1 10 - 11 1247 59
XM_017026885.1 XP_016882374.1 10 - 11 1250 59
XM_017026886.1 XP_016882375.1 11 - 12 2749 59
XM_017026887.1 XP_016882376.1 11 - 12 1219 59
XM_017026888.1 XP_016882377.1 11 - 12 1699 59
XM_017026889.1 XP_016882378.1 10 - 11 1241 59
XM_017026890.1 XP_016882379.1 10 - 11 1322 59
XM_017026891.1 XP_016882380.1 10 - 11 1199 59
XM_017026892.1 XP_016882381.1 10 - 11 1181 59
XM_017026893.1 XP_016882382.1 9 - 10 901 59
XM_017026894.1 XP_016882383.1 9 - 10 883 59
XM_017026895.1 XP_016882384.1 9 - 10 881 59
XM_017026896.1 XP_016882385.1 10 - 11 1247 59
GenBank mRNA BC006101.1 - 9 - 10 810 59
BT007438.1 - 9 - 10 756 59
M22596.1 - 1 - 2 42 59
M36542.1 - 9 - 10 810 59
M36653.1 - 9 - 10 870 59
M36718.1 - 9 - 10 801 59
X13809.1 - 9 - 10 801 59
X13810.1 - 9 - 10 810 59
X53468.1 - 9 - 10 870 59

View Full Product Details