The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020439.2 | NP_065172.1 | 3 - 4 | 328 | 64 |
XM_017001866.1 | XP_016857355.1 | 3 - 4 | 471 | 64 | |
XM_017001867.1 | XP_016857356.1 | 3 - 4 | 311 | 64 | |
GenBank mRNA | AF428261.1 | - | 3 - 4 | 298 | 64 |
AK095713.1 | - | 3 - 4 | 471 | 64 | |
AK289675.1 | - | 3 - 4 | 311 | 64 | |
AL049688.1 | - | 2 - 3 | 243 | 64 | |
AY212935.1 | - | 2 - 3 | 234 | 64 | |
BC032787.1 | - | 3 - 4 | 295 | 64 |