Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMS1
Entrez Gene ID
5378
Gene Name
PMS1 homolog 1, mismatch repair system component
Gene Aliases
HNPCC3, MLH2, PMSL1, hPMS1
Chromosome Location
Chr.2: 189784085 - 189877629 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000534.4 NP_000525.1 5 - 6 1115 112
NM_001128144.1 NP_001121616.1 5 - 6 1115 112
NM_001289408.1 NP_001276337.1 4 - 5 1012 112
NM_001289409.1 NP_001276338.1 3 - 4 829 112
NM_001321045.1 NP_001307974.1 6 - 7 1241 112
NM_001321046.1 NP_001307975.1 4 - 5 932 112
NM_001321047.1 NP_001307976.1 5 - 6 1292 112
NM_001321048.1 NP_001307977.1 6 - 7 1212 112
XM_011511356.2 XP_011509658.1 5 - 6 1401 112
XM_017004345.1 XP_016859834.1 4 - 5 549 112
XM_017004346.1 XP_016859835.1 4 - 5 512 112
XM_017004348.1 XP_016859837.1 5 - 6 841 112
XM_017004350.1 XP_016859839.1 4 - 5 541 112
XM_017004351.1 XP_016859840.1 3 - 4 503 112
GenBank mRNA AB102870.1 - 4 - 5 586 112
AB102876.1 - 4 - 5 586 112
AK295602.1 - 4 - 5 608 112
AK304634.1 - 4 - 5 451 112
AK308311.1 - 5 - 6 713 112
AK316215.1 - 3 - 4 349 112
BC036376.1 - 5 - 6 824 112
BC096330.1 - 5 - 6 656 112
BC096331.1 - 5 - 6 655 112
BC096332.1 - 5 - 6 656 112

View Full Product Details