The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009185.2 | NP_001009185.1 | 15 - 16 | 1614 | 84 |
NM_001205247.1 | NP_001192176.1 | 15 - 16 | 1509 | 84 | |
NM_001205248.1 | NP_001192177.1 | 15 - 16 | 1601 | 84 | |
NM_001205250.1 | NP_001192179.1 | 15 - 16 | 1611 | 84 | |
NM_001205251.1 | NP_001192180.1 | 14 - 15 | 1376 | 84 | |
NM_015256.3 | NP_056071.2 | 15 - 16 | 1614 | 84 | |
GenBank mRNA | AB020644.1 | - | 15 - 16 | 1574 | 84 |
AF129166.1 | - | 15 - 16 | 1544 | 84 | |
AK122702.1 | - | 4 - 5 | 1072 | 84 | |
AK294291.1 | - | 15 - 16 | 1484 | 84 | |
BC026161.1 | - | 15 - 16 | 1510 | 84 | |
BC047453.1 | - | 14 - 15 | 1298 | 84 | |
DQ083031.1 | - | 15 - 16 | 1475 | 84 |