Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
Chromosome Location
Chr.10: 8045421 - 8075201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002295.1 NP_001002295.1 4 - 5 1485 86
NM_002051.2 NP_002042.1 4 - 5 1482 86
XM_005252442.2 XP_005252499.1 4 - 5 1467 86
XM_005252443.4 XP_005252500.1 4 - 5 1392 86
GenBank mRNA BC003070.2 - 4 - 5 1376 86
BC006793.1 - 4 - 5 1487 86
M69106.1 - 3 - 4 1047 86
X55037.1 - 3 - 4 1049 86
X55122.1 - 3 - 4 1126 86
X58072.1 - 3 - 4 1076 86

View Full Product Details