Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SEPT8
- Entrez Gene ID
-
23176
- Gene Name
- septin 8
- Gene Aliases
-
SEP2
- Chromosome Location
-
Chr.5: 132750817 - 132778216 on Build GRCh38
View Full Product Details