The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176793.1 | NP_789763.1 | 4 - 5 | 70 | |
NM_176794.1 | NP_789764.1 | 5 - 6 | 70 | ||
GenBank mRNA | AF086122.1 | - | 3 - 4 | 70 | |
AW513604.1 | - | 1 - 2 | 171 | 70 | |
BC053373.1 | - | 4 - 5 | 70 | ||
BU072283.1 | - | 1 - 2 | 171 | 70 |