The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 2 - 3 | 466 | 81 |
NM_080599.2 | NP_542166.1 | 2 - 3 | 636 | 81 | |
XM_011519449.2 | XP_011517751.1 | 2 - 3 | 514 | 81 | |
GenBank mRNA | AB037829.1 | - | 1 - 2 | 835 | 81 |
AF301013.1 | - | 2 - 3 | 459 | 81 | |
AF318574.1 | - | 2 - 3 | 436 | 81 | |
AK096191.1 | - | 3 - 4 | 81 | ||
AY013249.1 | - | 1 - 2 | 385 | 81 | |
BC114964.1 | - | 2 - 3 | 430 | 81 | |
BC115737.1 | - | 2 - 3 | 430 | 81 | |
DA188353.1 | - | 2 - 3 | 466 | 81 |