The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282286.1 | NP_001269215.1 | 4 - 5 | 592 | 100 |
GenBank mRNA | AA524246.1 | - | 1 - 2 | 100 | |
AF263541.1 | - | 11 - 12 | 1388 | 100 | |
AK303744.1 | - | 4 - 5 | 560 | 100 | |
AL561586.3 | - | 4 - 5 | 592 | 100 | |
GU139739.1 | - | 2 - 3 | 299 | 100 | |
GU139744.1 | - | 3 - 4 | 462 | 100 | |
Y09305.1 | - | 4 - 5 | 486 | 100 |