The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017417.1 | NP_059113.1 | 8 - 9 | 1587 | 69 |
GenBank mRNA | AJ271385.1 | - | 8 - 9 | 1587 | 69 |
AJ505959.1 | - | 1 - 2 | 115 | 69 | |
BC140888.1 | - | 8 - 9 | 1553 | 69 | |
BC140889.1 | - | 8 - 9 | 1553 | 69 |