The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282485.1 | NP_001269414.1 | 3 - 4 | 818 | 65 |
NM_006259.2 | NP_006250.1 | 3 - 4 | 818 | 65 | |
XM_005263126.3 | XP_005263183.1 | 3 - 4 | 1253 | 65 | |
XM_017008413.1 | XP_016863902.1 | 3 - 4 | 1255 | 65 | |
XM_017008414.1 | XP_016863903.1 | 3 - 4 | 1250 | 65 | |
XM_017008415.1 | XP_016863904.1 | 3 - 4 | 660 | 65 | |
XM_017008416.1 | XP_016863905.1 | 3 - 4 | 1172 | 65 | |
XM_017008417.1 | XP_016863906.1 | 2 - 3 | 636 | 65 | |
XM_017008418.1 | XP_016863907.1 | 2 - 3 | 636 | 65 | |
GenBank mRNA | AK297673.1 | - | 3 - 4 | 765 | 65 |
D70899.1 | - | 2 - 3 | 623 | 65 | |
X94612.1 | - | 2 - 3 | 637 | 65 |