Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKG2
Entrez Gene ID
5593
Gene Name
protein kinase, cGMP-dependent, type II
Gene Aliases
PRKGR2, cGK2, cGKII
Chromosome Location
Chr.4: 81086874 - 81217837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282481.1 NP_001269410.1 2 - 3 130 76
NM_001282482.1 NP_001269411.1 2 - 3 130 76
NM_001282483.1 NP_001269412.1 2 - 3 130 76
NM_001282485.1 NP_001269414.1 7 - 8 1181 76
NM_006259.2 NP_006250.1 7 - 8 1181 76
XM_005263126.3 XP_005263183.1 7 - 8 1616 76
XM_017008413.1 XP_016863902.1 7 - 8 1618 76
XM_017008414.1 XP_016863903.1 7 - 8 1613 76
XM_017008415.1 XP_016863904.1 7 - 8 1023 76
XM_017008416.1 XP_016863905.1 7 - 8 1535 76
XM_017008417.1 XP_016863906.1 6 - 7 999 76
XM_017008418.1 XP_016863907.1 6 - 7 999 76
GenBank mRNA AK296979.1 - 2 - 3 117 76
AK297673.1 - 7 - 8 1128 76
D70899.1 - 6 - 7 986 76
DA462272.1 - 2 - 3 130 76
X94612.1 - 6 - 7 1000 76

View Full Product Details