The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007022.2 | NP_001007023.2 | 14 - 15 | 1883 | 71 |
XM_005271054.3 | XP_005271111.1 | 13 - 14 | 1801 | 71 | |
XM_011541815.2 | XP_011540117.1 | 14 - 15 | 1888 | 71 | |
XM_011541819.2 | XP_011540121.1 | 14 - 15 | 1888 | 71 | |
XM_017001876.1 | XP_016857365.1 | 14 - 15 | 1680 | 71 | |
XM_017001877.1 | XP_016857366.1 | 13 - 14 | 1593 | 71 | |
XM_017001878.1 | XP_016857367.1 | 13 - 14 | 1771 | 71 | |
XM_017001879.1 | XP_016857368.1 | 13 - 14 | 1801 | 71 | |
XM_017001880.1 | XP_016857369.1 | 12 - 13 | 1684 | 71 | |
XM_017001881.1 | XP_016857370.1 | 11 - 12 | 1556 | 71 | |
XM_017001882.1 | XP_016857371.1 | 7 - 8 | 2001 | 71 | |
GenBank mRNA | AB033055.1 | - | 14 - 15 | 1803 | 71 |
BM971230.1 | - | 4 - 5 | 71 | ||
BQ894001.1 | - | - | 550 | 71 |