The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321978.1 | NP_001308907.1 | 3 - 4 | 521 | 74 |
NM_001321979.1 | NP_001308908.1 | 3 - 4 | 521 | 74 | |
NM_014317.4 | NP_055132.2 | 3 - 4 | 521 | 74 | |
GenBank mRNA | AB209763.1 | - | 2 - 3 | 2559 | 74 |
AB210838.1 | - | 3 - 4 | 279 | 74 | |
AK024802.1 | - | 3 - 4 | 286 | 74 | |
AK223414.1 | - | 3 - 4 | 286 | 74 | |
BC049211.1 | - | 3 - 4 | 231 | 74 | |
BC063635.1 | - | 3 - 4 | 249 | 74 |